Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN394

Sigma-Aldrich

Anti-PSMD9 Antibody, clone 16C5.1

clone 16C5.1, from mouse

Synonym(s):

26S proteasome non-ATPase regulatory subunit 9, 26S proteasome regulatory subunit p27, Homolog of rat Bridge 1, Proteasome macropain non-ATPase subunit 9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$322.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16C5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PSMD9(5715)

General description

26S proteasome non-ATPase regulatory subunit 9 (UniProt O00233; also known as 26S proteasome regulatory subunit p27, Homolog of rat Bridge 1, Proteasome macropain non-ATPase subunit 9) is encoded by the PSMD9 gene (Gene ID 5715) in human. PSMD9 is a proteasomal assembly chaperone that forms a stable subcomplex with two AAA-ATPases, PSMC3 and PSMC6, in assisting the assembly of the 20S and 19S particles to form the holo complex. PSMD9 contains a PDZ-like domain (a.a. 108-195) that is reported to interact with a short 4- to 7-amino acid linear sequence motifs (SLIMs) at the C-terminal end of its client proteins, including the RNA-binding protein hnRNPA1, the ribosomal protein S14, the growth hormone CSH1, the transcription factor E12, and the IL6 receptor. PSMD9 and hnRNPA1 interaction is found to play a role in IκBα proteasomal degradation, where hnRNPA1 functions as shuttle receptor that recruits IκBα to the proteasome for degradation.

Specificity

Expected to react with spliced isoforms p27-L and p27-S, but not spliced isoform 3.

Immunogen

Epitope: Internal.
GST-tagged recombinant human PSMD9 internal fragment.

Application

Anti-PSMD9 Antibody, clone 16C5.1 is an antibody against PSMD9 for use in Western Blotting, Immunohistochemistry (Paraffin).
Research Category
Neuroscience
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected PSMD9 in 10 µg of HepG2 cell lysate.
Immunohistochemistry Analysis: An 1:50 dilution from a representative lot detected PSMD9 in normal human kidney and prostate tissues, as well as human prostate cancer tissue sections.

Quality

Evaluated by Western Blotting in SHSY-5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PSMD9 in 10 µg of SHSY-5Y cell lysate.

Target description

~26 kDa observed. 24.68 kDa (isoforms p27-L) and 22.76 kDa (isoforms p27-S) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service