Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN376

Sigma-Aldrich

Anti-Alanyl-tRNA synthetase (AARS Antibody), clone M6-P2E5

clone P2E5, 1 mg/mL, from mouse

Synonym(s):

Alanine-tRNA ligase, cytoplasmic, Alanyl-tRNA synthetase, AlaRS, Renal carcinoma antigen NY-REN-42

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$333.00

$333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$333.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$333.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P2E5, monoclonal

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AARS1(16)

General description

Alanyl-tRNA synthetase (AARS), also known as Alanine-tRNA ligase, AlaRS, or Renal carcinoma antigen NY-REN-42, and encoded by the gene AARS, is a key transfer RNA synthetase responsible for the attachment of alanine to the tRNA(Ala) for protein translation. Alanine-tRNA ligase also can edit incorrectly charged tRNA(Ala) for proper translation. Alanine-tRNA ligase is a monomer and is localized in the cytoplasm and ubiquitously expressed. Mutations in Alanine-tRNA ligase have been linked to a progressive peripheral nervous disorder called Charcot-Marie Tooth disease. Moreover autoantibodies against aminoacyl tRNA synthetases are some of the most common autoantibodies detected in autoimmune diseases with nearly 25-35% of patients having them, and some of these such as autoantibodies to Alanine-tRNA ligase have also been associated with autologous antibodies in renal carcinoma patients.

Immunogen

Linear peptide corresponding to the C-terminus of human Alanyl-tRNA synthetase (AARS).

Application

This Anti-Alanyl-tRNA synthetase (AARS Antibody), clone M6-P2E5 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Alanyl-tRNA synthetase (AARS).
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Alanyl-tRNA synthetase (AARS) in 10 µg of HepG2 cell lysate.
Immunohistochemistry Analysis: A representative lot detected Alanyl-tRNA synthetase (AARS) in human breast cancer tissue (Prof. Ayham Alnabulsi, University of Aberdeen)

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Alanyl-tRNA synthetase (AARS) in 10 µg of HeLa cell lysate.

Target description

~110 kDa observed

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service