Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN2185

Sigma-Aldrich

Anti-LGMN Antibody, clone 6A7.2

clone 6A7.2, from mouse

Synonym(s):

Asparaginyl endopeptidase, Protease cysteine 1, Legumain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

6A7.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LGMN(5641)

General description

Legumain (UniProt: Q99538; EC:3.4.22.34, also known as Asparaginyl endopeptidase; Protease, cysteine 1) is encoded by LGMN (also known as PRSC1) gene (Gene ID 5641) in human. Legumain is a lysosomal enzyme with strict specificity of the hydrolysis of asparaginyl bonds. It is shown to be ubiquitously distributed with abundance in kidney, heart, and placenta. In the zymogen form, the uncleaved propeptide blocks access to the active site. It is activated autocatlytically by acidic pH and the active form is shown to be monomeric. Legumain plays a pivotal role in the endosomal/lysosomal degradation system and is involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation. Legumain deficiency causes the accumulation of pro-cathepsins B, H and L. Overexpression of legumain is associated with invasion and metastasis in a majority of solid tumors.

Specificity

Clone 6A7.2 of detects LGMN in multiple human tissues by Western blotting and immunohistochemistry.

Immunogen

GST-tagged recombinant fragment corresponding to 95 amino acids from the internal region of human LGMN.

Application

Anti-LGMN, clone 6A7.2 Antibody, Cat. No. MABN2185, is a highly specific mouse monoclonal antibody that targets LGMN and has been tested in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LGMN in human placenta, human kidney, and human small intestine.


Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected LGMN in 10 µg of human heart tissue lysate.

Target description

~ 55 kDa observed; 49.4 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service