MABN1835
Anti-DYRK1A Antibody, clone 7F3
clone 7F3, from mouse
Synonym(s):
Dual specificity tyrosine-phosphorylation-regulated kinase 1A
Select a Size
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
7F3, monoclonal
species reactivity
human, rat, monkey
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... DYRK1A(1859)
rat ... Dyrk1A(25255)
rhesus monkey ... Dyrk1A(693812)
General description
Specificity
Immunogen
Application
Immunocytochemistry Analysis: A representative lot, pre-conjugated with Alexa Fluor™ 568, immunostained the soma and processes of 2% formaldehyde-fixed, 0.05% saponin-permeabilized E-18 rat hippocampal neurons by fluorescent immunocytochemistry. DYRK1A immunoreactivity was more abundant in nuclei than in the cytoplasm. Co-localization of DYRK1A and clathrin heavy chain (CHC) immunoreactivity was very prominent in the processes and less significant in the soma (Murakami, N., et al. (2012). PLoS One. 7(4):e34845).
Immunofluorescence Analysis: A representative lot detected DYRK1A cellular localization in GFAP-positive astrocytes and in vimentin-positive endothelial cells by dual fluorescent immunohistochemistry staining of 10% formalin-fixed, PEG 1000-embeded human brain tissue sections (Wegiel, J., et al. (2004). Brain Res. 1010(1-2):69-80).
Immunohistochemistry Analysis: A representative lot detected DYRK1A immunoreactivity in paraffin-embedded fetal (20-week gestation) Down syndrome (DS) white matter associated with structures resembling the omnipresent cytoskeleton filamentous network in axons (Dowjat, K., et al. (2012). J. Neuropathol. Exp. Neurol. 71(12):1100-1112).
Immunohistochemistry Analysis: A representative lot detected age- and brain region-dependent DYRK1A immunoreactivity in 10% formalin-fixed, PEG 1000-embeded human brain tissue sections (Wegiel, J., et al. (2004). Brain Res. 1010(1-2):69-80).
Western Blotting Analysis: A representative lot detected both endogenous monkey DYRK1A and exogenously expressed full-length rat DYRK1A in COS-7 cells, as well as DYRK1A in human hippocampus tissue extracts. Antibody blocking with the immunogen prior to Western blotting abolished target band detection (Wegiel, J., et al. (2004). Brain Res. 1010(1-2):69-80).
Neuroscience
Quality
Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected DYRK1A in rat cerebellum tissue section.
Target description
Physical form
Storage and Stability
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service