Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABN1772

Sigma-Aldrich

Anti-pan Thyroid Hormone Receptor Antibody, clone N403/63

clone N403/63, from mouse

Synonym(s):

Thyroid hormone receptor alpha, Nuclear receptor subfamily 1 group A member 1, V-erbA-related protein 7, EAR-7, c-erbA-1, c-erbA-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N403/63, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THRA(7067)

General description

Thyroid hormone receptor alpha (UniProt P10827; also known as Nuclear receptor subfamily 1 group A member 1, V-erbA-related protein 7, EAR-7, c-erbA-1, c-erbA-alpha) is encoded by the THRA (also known as CHNG6, EAR7, ERBA, ERBA1, NR1A1, THRA1, THRA2) gene (Gene ID 7067) in human. Thyroid hormone receptors (THRs) bind to short, repeated sequences of DNA known as T3 response elements (TREs) and mediate thyroid hormone (TH) regulated gene transcriptions. THRs belong to the super family of single chain nuclear receptors with similar folded domain structures, including an N-terminal domain (A–B domain), a central DNA-binding domain (DBD), and a C-terminal ligand-binding domain (LBD). The LBD dictates ligand specificity and modulates homodimerization or heterodimerization with other family members. The DBDs of all THRs are highly homologous and each of these proteins can form heterodimers and homodimers. All THRs contain activation 1 (AF-1) region in the A–B domain; THRα1 and THRβ1-3 also contain a C-terminal AF-2 domain. THRα1 and THRα2 are truncated splice variants that do not contain AF-2 and do not bind the active TH form T3. In the absence of T3, THR recruits a co-repressor, such as N-CoR or N-CoR2.46, to the TRE sites, resulting in target transcription suppression as a result of co-repressor-mediated chromatin epigenetic modification. Upon T3 binding, the co-repressors are released, leading to the exposure of the AF-2 domain and recruitment of co-activators, such as SRC-1, that induce gene transcriptions by inducing chromatin remodeling and alter the DNA conformation.

Specificity

Exprected to react with all four spliced isoforms of human thyroid hormone receptor alpha reported by UniProt (P10827). Clone N403/63 also cross-reacts with both spliced isoforms of human thyroid hormone receptor beta (UniProt P10828), but not potassium channel Kv2.1.

Immunogen

Recombinant full-length human thyroid hormone receptor Alpha-1 isoform.

Application

Detect pan Thyroid Hormone Receptor using this Anti-pan Thyroid Hormone Receptor Antibody, clone N403/63 validated for use in Immunohistochemistry (Paraffin), Western Blotting.
Research Category
Neuroscience
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected thyroid hormone receptor in 10 µg of human and rat brain tissue lysates.
Immunohistochemistry Analysis: An 1:250 dilution from a representative lot detected thyroid hormone receptor in human cerebral cortex, mouse cerebellum, and rat kidney tissue.
Western Blotting Analysis: A representative lot detected the transient expression of c-Myc-tagged human thyroid hormone receptor alpha (isoform Alpha-1) and beta (isoforms Beta-1 & Beta-2) construts in transfected COS-7 cells (Courtesy of Professor. James S. Trimmer, UC Davis, CA, USA).
Immunohistochemistry Analysis: A representative lot detected thyroid hormone receptor immunoreactivity in rat whole brain sections (Courtesy of Professor. James S. Trimmer, UC Davis, CA, USA).

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Thyroid Hormone Receptor in 10 µg of mouse brain tissue lysate.

Target description

~70 kDa observed. Target band appears larger than the calculated molecular weights of 46.81 kDa (isoform Alpha-1), 54.81 kDa (isoform Alpha-2), 50.75 kDa (isoform Alpha-3), 50.47 kDa (isoform Alpha-4), 52.79 kDa (isoform Bet-1), 54.45 kDa (isoform Beta-2).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service