Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN1729

Sigma-Aldrich

Anti-Ankyrin-B Antibody, clone N105/13

clone N105/13, from mouse

Synonym(s):

Ankyrin-2, ANK-2, Ankyrin-B, Brain ankyrin, Brank-2, Non-erythroid ankyrin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N105/13, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANK2(287)

General description

Ankyrin-2 (UniProt Q01484; also known as ANK-2, Ankyrin-B, Brain ankyrin, Brank-2, Non-erythroid ankyrin) is encoded by the ANK2 (also known as LQT4) gene (Gene ID 287) in human. Ankyrins are scaffold proteins that play essential roles in providing plasma membrane with mechanical support and in controlling excitable tissues by connecting numerous ion channels, cell adhesion molecules, and receptors to the spectrin-based cytoskeleton. There are three human ankyrins (ankyrin-R, -B, and -G encoded by ANK1, ANK2, and ANK3 gene, respectively), all share similar domain organization, including an N-terminal membrane-binding domain composed of 24 ankyrin (ANK) repeats, a spectrin-binding region comprised of two ZU5 domains, a UPA domain, a death domain (DD) and a variable C-terminal regulatory domain. The three ankyrins, however, do have distinct and non-overlapping functions in specific membrane domains coordinated by ankyrin-spectrin networks. Ankyrins mediate a wide range of physiological functions by interacting with and anchoring numerous membrane targets to specific membrane micro-domains in coordination with spectrin-based cytoskeletal structures. Examples include AnkB-mediated coordination of voltage-gated sodium channels, Na+/K+- ATPase, Na+/Ca2+ exchanger, and inositol 1,4,5-triphosphate receptors in cardiomyocytes, as well as the dystrophin/dystroglycan complex in skeletal muscles. In addition, multiple ankyrin isoforms generated through alternative splicing of a given ANK gene can further impart functional specificity and distinct subcellular distribution.

Specificity

Clone N105/13 detects human, mouse, and rat Ankyrin-2/Ankyrin-B, but not Ankyrin-3/Ankyrin-G. Expected to react with spliced isoform 2 (Q01484-2), isoform 3 (Q01484-4), and isoform 4 (Q01484-5), but not isoform 5 (Q01484-7) reported by UniProt.

Immunogen

Epitope: Ankyrin repeats 6-14.
Recombinant protein corresponding to Ankyrin repeats 6-14 of human Ankyrin-B.

Application

Detect Ankyrin-B using this Anti-Ankyrin-B Antibody, clone N105/13 validated for use in Western Blotting, Immunohistochemistry.
Research Category
Neuroscience
Research Sub Category
Adhesion (CAMs)
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Ankyrin-B in 10 µg of human brain tissue lysate.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Ankyrin-B in human cerebellum, human cerebral cortex, mouse cerebellum, mouse cerebral cortex, and rat brain tissue sections.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Ankyrin-B in 10 µg of mouse brain tissue lysate.

Target description

>200 kDa observed. 205.8 kDa (isoform 2), 433.7 kDa (isoform 3), and 204.8 kDa (isoform 4) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service