Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN1725

Sigma-Aldrich

Anti-Thyroid Hormone Receptor β-1 Antibody, clone N402/46

clone N402/46, from mouse

Synonym(s):

Thyroid hormone receptor beta, Nuclear receptor subfamily 1 group A member 2, c-erbA-2, c-erbA-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N402/46, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... THRB(7068)

General description

Thyroid hormone receptor beta (UniProt: P10828; also known as Nuclear receptor subfamily 1 group A member 2, c-erbA-2, c-erbA-beta) is encoded by the THRB (also known as ERBA2, NR1A2, THR1) gene (Gene ID: 7068) in human. Thyroid hormone receptors (TRs) are ligand dependent members of the steroid/retinoic acid superfamily of transcription factors. Thyroid hormone receptor beta is a nuclear hormone receptor that has a transactivation domain at its N-terminal, a ligand-binding domain at the C-terminal, and a DNA-binding domain that binds to sequences of promoter DNA known as hormone response elements. It serves as a high affinity receptor for thyroid hormones, including triiodothyronine (T3) and thyroxine and serves as a hormone-dependent transcriptional regulator that binds DNA sequences and is activated by T3. It plays an important role in vertebrate development, specifically in developing limbs and face. 9-cis-retinoic acid receptor (RXR) and retinoic acid receptor (RAR) receptor families are often associated with thyroid hormone receptor -1 and form homodimers and heterodimers. Mutations in this gene are known to be a cause of generalized thyroid hormone resistance (GTHR), a syndrome characterized by goiter and high levels of circulating thyroid hormone (T3-T4), with normal or slightly elevated thyroid stimulating hormone (TSH).

Specificity

Clone N402/46 detects Thyroid Hormone Receptor beta-1 in multiple human tissues.

Immunogen

A full-length human recombinant Thyroid Hormone Receptor beta-1.

Application

Anti-Thyroid Hormone Receptor -1, clone N402/46 Antibody, Cat. No. MABN1725, is a highly specific mouse monoclonal antibody that targets Thyroid Hormone Receptor -1 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Thyroid Hormone Receptor -1 in human pancreas and human liver tissue.

Western Blotting Analysis: A representative lot detected Thyroid Hormone Receptor beta-1 in COS cells transiently transfected with Myc-tagged TRalpha1, Myc-tagged TRbeta1, Myc-tagged TRbeta2 or untagged Kv2.1 plasmid. (Courtesy of James Trimmer, Ph.D., University of California at Davis, USA).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in human cerebellum tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Thyroid Hormone Receptor -1 in human cerebellum tissue.

Target description

~60 kDa observed; 52.79 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service