Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1593

Sigma-Aldrich

Anti-DGCR8 Antibody, clone 11C10.1

clone 11C10.1, from mouse

Synonym(s):

Microprocessor complex subunit DGCR8, DiGeorge syndrome critical region 8, Pasha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11C10.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DGCR8(54487)

General description

Microprocessor complex subunit DGCR8 (UniProt Q8WYQ5; also known as DiGeorge syndrome critical region 8, Pasha) is encoded by the DGCR8 (also known as DGCRK6, LP4941) gene (ORF C22orf12; Gene ID 54487) in human. The DGCR8 gene encodes a double-stranded RNA-binding protein that functions as a non-catalytic subunit of the microprocessor complex which mediates the biogenesis of microRNAs (miRNA) from the primary microRNA (pri-microRNA) transcript. miRNAs are initially transcribed as pri-microRNA with a local stem-loop that is cleaved by the microprocessor complex to produce the precursor microRNA (pre-miRNA). The microprocessor complex is comprised of the RNase Drosha and the RNA-binding protein DGCR8. DGCR8 is reported to be phosphorylated by Erk and Abl, an event that promotes DGCR8 stability and stimulates Drosha recruitment and subsequent substrate pri-microRNAs processing.

Specificity

Expected to react with spliced isoforms 1 and 3, but not isoform 2 of human DGCR8 reported by UniProt (Q8WYQ5).

Immunogen

Epitope: Region between WW and DRBM 1 domain.
GST-tagged recombinant fragment from the region between the WW and DRBM 1 domain of human DGCR8.

application

Anti-DGCR8 Antibody, clone 11C10.1 is an antibody against DGCR8 for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected DGCR8 in 10 µg of HeLa cell lysate.

Target description

~120 kDa observed. 86.05 kDa (isoform 1) and 82.75 kDa (isoform 3). The detected band size is consistent with the reported apparent molecular weight (Ha, M., and Kim, V.N. (2014). Nat. Rev. Mol. Cell Biol. 15(8):509-524).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service