Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1556

Sigma-Aldrich

Anti-GGA3 Antibody, clone 3G5.1

clone 3G5.1, from mouse

Synonym(s):

ADP-ribosylation factor-binding protein GGA3, Golgi-localized gamma ear-containing ARF-binding protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G5.1, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GGA3(23163)

General description

ADP-ribosylation factor-binding protein GGA3 (UniProt: Q9NZ52; also known as Golgi-localized, gamma ear-containing, ARF-binding protein 3) is encoded by the GGA3 (also known as KIAA0154) gene (Gene ID: 23163) in human. GGA3 is a monomeric, ubiquitously expressed protein found in trans-Golgi network (TGN) membrane and endosome membrane. It plays a vital role in protein sorting and trafficking between the trans-Golgi network and endosomes. It mediates the ARF-dependent recruitment of clathrin to the TGN and binds ubiquitinated proteins and membrane cargo molecules with a cytosolic acidic cluster-dileucine (AC-LL) motif. Four isoforms of GGA3 have been described that are produced by alternative splicing. It contains a VHS domain (aa 16-146) that functions as a recognition module for sorting signals composed of an acidic cluster followed by two leucines. Its GAT domain (aa 171-298) is responsible for interaction with ARF-GTP, UBC, and RAB GTPase Binding Effector Protein 1 (RABEP1) and prevents ARF-GTP hydrolysis. GGA3 also has a GAE domain (aa 594-715) that binds accessory proteins regulating GGAs function. GGA3 also contains an auto-inhibitory motif (aa 391-395). GGA3 can be phosphorylated by casein kinase 2 and dephosphorylated by protein phosphatase 2A. GGA3 phosphorylation allows the internal AC-LL motif to bind VHS domain and inhibit the recognition of cargo signals.

Specificity

Clone 3G5.1 detects ADP-ribosylation factor-binding protein GGA3 in human and murine cells. It targets an epitope within 103 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 103 amino acids from the internal region of human ADP-ribosylation factor-binding protein GGA3.

Application

Anti-GGA3, clone 3G5.1, Cat. No. MABN1556, is a highly specific mouse monoclonal antibody that targets ADP-ribosylation factor-binding protein GGA3 and has been tested for use in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in Neuro-2a cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GGA3 in 10 µg of Neuro-2a cell lysate.

Target description

~80 kDa observed; 78.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service