Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN1122

Sigma-Aldrich

Anti-PSMA7, clone EPR5836, Rabbit Monoclonal Antibody

clone EPR5836, from rabbit

Synonym(s):

Proteasome subunit alpha type-7, Proteasome subunit RC6-1, Proteasome subunit XAPC7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR5836, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PSMA7(5688)

General description

PSMA (prostate specific membrane antigen) is a type II, integral membrane glycoprotein, composed of a 19 amino acid intracellular domain containing the N-terminus, a 24 amino acid transmembrane region, and a 707 amino acid extracellular C-terminal domain. The PSMA gene has been cloned and sequenced, and has been localized to chromosome 11q. PSMA is highly expressed in prostate secretory-acinar epithelium, in some benign extraprostatic epithelial cells from breast, duodenum, and kidney tissues, and in prostate cancer. Recently, evidence of limited and specific endothelial PSMA expression has been discovered: the neovasculature of a wide variety of malignant neoplasms (lung, colon, breast, etc.) demonstrates PSMA expression. This finding suggests that PSMA may be an effective target for monoclonal antibody-based anti-neovasculature therapy. PSMA has also been identified in the human nervous system. Based on activity, immunoreactivity, and mRNA sequence comparison with one form of NAALADase, a neuropeptide that may modulate glutaminergic transmission in the nervous system, PSMA was found to be indistinguishable from NAALADase derived from human cerebellar cell isolates. Therefore, the form of NAALADase also known as PSMA is expressed in brain, where it makes up a significant fraction of brain NAALADase activity.

Immunogen

Synthetic peptide corresponding to human PSMA7.

Application

Immunohistochemistry Analysis: A 1:250-500 dilution from a representative lot detected PSMA7 in human breast carcinoma tissue.

Produced in collaboration with Epitomics
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-PSMA7, clone EPR5836, Rabbit Monoclonal Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of PSMA7.

Quality

Evaluated by Western Blotting in A549, MCF-7, HEK293, and HeLa cell lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected PSMA7 in A549, MCF-7, HEK293, and HeLa cell lysates.

Target description

~27 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service