Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1117

Sigma-Aldrich

Anti-Pannexin 1, clone EPR5556, Rabbit Monoclonal Antibody

clone EPR5556, from rabbit

Synonym(s):

Pannexin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR5556, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PANX1(24145)

General description

Pannexin 1 (PANX1) belongs to the innexin family. Innexin family members are the structural components of gap junctions. Pannexin 1 and pannexin 2 are abundantly expressed in central nerve system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that Pannexin 1 alone and in combination with pannexin 2 may form cell type-specific gap junctions with distinct properties.

Immunogen

Synthetic peptide corresponding to human Pannexin 1.

Application

Produced in collaboration with Epitomics
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-Pannexin 1, clone EPR5556, Rabbit Monoclonal Antibody is validated for use in Western Blotting for the detection of Pannexin 1.

Quality

Evaluated by Western Blotting in human fetal brain, SH-SY5Y, Caco-2, and NIH:OVACAR-3 lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected Pannexin 1 in human fetal brain, SH-SY5Y, Caco-2, and NIH:OVACAR-3 lysates.

Target description

~48 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service