Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF980

Sigma-Aldrich

Anti-CD22 Antibody, clone CY34

clone CY34, from mouse

Synonym(s):

B-cell receptor CD22, B-lymphocyte cell adhesion molecule, BL-CAM, Sialic acid-binding Ig-like lectin 2, Siglec-2, T-cell surface antigen Leu-14, CD22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CY34, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd22(12483)

General description

B-cell receptor CD22 (UniProt P20273; also known as B-lymphocyte cell adhesion molecule, BL-CAM, Sialic acid-binding Ig-like lectin 2, Siglec-2, T-cell surface antigen Leu-14) is encoded by the Cd22 (also known as A530093D23, Lyb-8, Lyb-8a, Lyb8, Siglec2) gene (Gene ID 12483) in murine species. Murine CD22 (mCD22) is a B cell-associated adhesion protein with seven extracellular Ig-like domains that has 62% amino acid homology to its human orthologue. CD22 is expressed at multiple stages during B cell ontogeny. Cytoplasmic hCD22 can be detected at the pro-B and pre-B, whereas expression of surface hCD22 tightly correlates with that of surface immunoglobulins (sIg), especially that of sIgD. Expression of hCD22 ceases upon B cell activation and terminal differentiation into plasma cells. Only the CD22+ subpopulation among sIgM+ tonsillar B cells can be stimulated by sIg cross-linking to induce intracellular calcium, which can be further enhanced by anti-CD22 treatment prior to sIg cross-linking. Crosstalk between CD22 and sIgM is also supported by the finding that CD22 can associate with sIgM and cross-linking of sIg can stimulate rapid tyrosine phosphorylation of CD22.

Specificity

Murine Cd22 gene is also known as Lyb-8. The epitope detected by clone CY34 is not expressed by all strains of mice due to Cd22 polymorphisms. Clone CY34 is known to react with Cd22 expressed on splenocytes from murine strains B10.D2 and BALB/c, C57BL/6, but not DBA/2J. Consequently, the allelic form of Cd22 from DBA/2J is designated as mCD22.1/Lyb-8.1 to distinquish it from the form of Cd22 recognized by clone CY34, originally designated as Lyb-8.2 (PMID 8100843 & 6984010).

Immunogen

Epitope: Extracellular Ig-like domains 1 & 2
Splenocytes from mouse strain B10.D2

Application

Flow Cytometry Analysis: A representative lot detected the exogenously expressed mCD22, but not mCD40, on the surface of respective BHK transfectants (Law, C.L., et al. (1993). J. Immunol. 151(1):175-187).
Flow Cytometry Analysis: A representative lot selectively detected the Lyb-8.2 allelic form of Cd22 on sIgM-positive B lymphocytes from BALB/c mice, but not the Lyb-8.1 allelic form of Cd22 expressed on B lymophocytes from DBA/2J mice (Law, C.L., et al. (1993). J. Immunol. 151(1):175-187).
Immunoprecipitation Analysis: A representative lot immunoprecipitated the Lyb-8.2 allelic form of Cd22 from the splenocytes of B10.D2 mouse strain (Symington F.W., et al. (1982). Immunogenetics. 16(5):381-391).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-CD22 Antibody, clone CY34 is validated for use in Flow Cytometry and Immunoprecipitation for the detection of CD22.

Quality

Evaluated by Flow Cytometry in splenocytes from mouse strain C57BL/6.

Flow Cytometry Analysis: 1.0 µg of this antibody detected CD22 on splenocytes from mouse strain C57BL/6.

Target description

~95 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service