Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF725

Sigma-Aldrich

Anti-CD86 (B7-2) Antibody (mouse), FITC, clone GL-1

clone GL-1 (GL1), 0.5 mg/mL, from rat

Synonym(s):

T-lymphocyte activation antigen CD86, Activation B7-2 antigen, Early T-cell costimulatory molecule 1, ETC-1, CD86

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

rat

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

GL-1 (GL1), monoclonal

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd86(12524)

General description

The GL-1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response. The GL-1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This rat monoclonal antibody Anti-CD86 (B7-2) (mouse), FITC, clone GL-1 validated for use in IHC, Immunofluorescence & Flow Cytometry for the detection of CD86 (B7-2).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 µg of this antibody detected CD86 (B7-2) in one million C57Bl/6 splenocytes.

Physical form

Purified Rat Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service