MABF447
Anti-CD45R (B220) Antibody (human/mouse), FITC, clone RA3-6B2
clone RA3-6B2, 0.5 mg/mL, from rat
Synonym(s):
Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, Lymphocyte antigen 5, Ly-5, T200, CD45
About This Item
biological source
rat
conjugate
FITC conjugate
antibody form
purified antibody
antibody product type
primary antibodies
clone
RA3-6B2, monoclonal
species reactivity
human, mouse
concentration
0.5 mg/mL
technique(s)
flow cytometry: suitable
isotype
IgG2aκ
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PTPRC(5788)
mouse ... Ptprc(19264)
General description
Immunogen
Application
Inflammation & Immunology
Quality
Flow Cytometry Analysis: 0.25 µg of this antibody detected CD45R (B220) in one million C57Bl/6 splenocytes.
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service