MABF358
Anti-CD45RO (human) Antibody, PE/Cy5, clone UCHL1
clone UCHL1, from mouse, PE-Cy5
Synonym(s):
Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, T200, CD45, CD45RO
About This Item
biological source
mouse
conjugate
PE-Cy5
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
UCHL1, monoclonal
species reactivity
human
technique(s)
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2aκ
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PTPRC(5788)
General description
Specificity
Immunogen
Application
Inflammation & Immunology
Cytokines & Cytokine Receptors
Immunohistochemistry Analysis: A representative lot detected human CD45RO in Immunohistochemistry. (Yamada M, et al.1996.;Sakkas LI, et al. 1998.)
Immunoprecipitation Analysis: A representative lot detected human CD45RO in Immunoprecipitation. (Ponsford M, et al. 2001.)
Quality
Flow Cytometry Analysis: 5 μL of this antibody detected CD45RO in one million human peripheral blood lymphocytes.
Target description
Linkage
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service