Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF345

Sigma-Aldrich

Anti-LAP (TGF-β1) (mouse) Antibody, APC, clone TW7-16B4

clone TW7-16B4, from mouse, Allophycocyanin conjugate

Synonym(s):

Transforming growth factor beta-1, TGF-beta-1, Latency-associated peptide, LAP, TGFB1, TGFB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

conjugate

Allophycocyanin conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

TW7-16B4, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Tgfb1(21803)

General description

LAP, also known as TGF-beta1 or TGFB1, belongs to the TGF-beta family. TGFB1 is a multifunctional protein that controls proliferation, differentiation and other functions in various cell types. TGFB1 is synthesized by many cells that have specific receptors for it. It positively and negatively regulates many other growth factors. LAP TGF-beta1 plays an important role in bone remodeling as it is a potent stimulator of osteoblastic bone formation, causing chemotaxis, proliferation and differentiation in committed osteoblasts. The ′straitjacket′ and ′arm′ domains encircle the growth factor monomers and are fastened together by strong bonding between Lys-56 and Tyr-103/Tyr-104. Activation of TGF-beta1 requires the binding of integrin alpha-V to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-beta binding proteins. The physical force unfastens the straitjacket and releases the active growth factor dimer.

Specificity

This antibody recognizes recombinant LAP, latent TGF-β, and pro-TGF-β.

Immunogen

Mouse Tgfb1-transduced P3U1 cells

Application

Detect TGFβ using this mouse monoclonal antibody, Anti-LAP (TGF-β1) (mouse) Antibody, APC, clone TW7-16B4 validated for use in Flow Cytometry, western blotting, IP & Neutralizing.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western Blotting Analysis: A representative lot detected human LAP (TGF-β1) in Western Blotting. (Oida T, et al. 2010.)

Immunoprecipitation Analysis: A representative lot detected human LAP (TGF-β1) in Immunoprecipitation. (Oida T, et al. 2010.)

Neutralization Assay Analysis: A representative lot detected human LAP (TGF-β1) in Neutralization Assay. (Oida T, et al. 2011.)

Quality

The antibody was purified by affinity chromatography, and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.

Flow Cytometry Analysis: 0.5 μg of this antibody detected LAP (TGF-β1) in one million CD3+CD28+IL-2-stimulated C57BL/6 mouse splenocytes.

Target description

44 kDa calculated

Physical form

Affinity
Purified mouse conjugate antibody in buffer containing PBS (pH 7.2) with up to 0.1% sodium azide.

Storage and Stability

The antibody solution should be stored undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service