Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF284

Sigma-Aldrich

Anti-RIG1 Antibody, clone 42

clone 42, from mouse

Synonym(s):

Retinoic acid receptor responder protein 3, HRAS-like suppressor 4, HRSL4, RAR-responsive protein TIG3, Retinoid-inducible gene 1 protein, Tazarotene-induced gene 3 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$401.00

$401.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$401.00


Estimated to ship onMay 02, 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

42, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLAAT4(5920)

General description

RIG1 is a cytoplasmically bound enzyme that exhibits phospholipase 1/2 activity and catalyzes the hydrolysis of various important membrane lipids including phosphatidylcholine and phosphatidylethanolamine containing lipids. RIG1 mimics PLA1/2 activities in many tissues and is widely expressed. Recent research has shown that RIG1 also functions as a cytoplasmic pattern receptor that detects viral RNA and acts as a cellular sensor for viral infection, which in turn can activate the proinflammatory response and the production of type-1 interferons against the infection.

Immunogen

Recombinant protein corresponding to human RIG1.

Application

Flow Cytometry Analysis: 0.25 µg of this antibody from a representative lot detected RIG1 in 1X10E6 THP-1 cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-RIG1 Antibody, clone 42 is validated for use in Western Blotting and Flow Cytometry for the detection of RIG1.

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected RIG1 in 10 µg of THP-1 cell lysate.

Target description

~17 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service