MABF2122
Anti-PCBP2 (hnRNP E2) Antibody, clone PC81-1
clone PC 81-1, from mouse
Synonym(s):
Poly(rC)-binding protein 2, Alpha-CP2, Heterogeneous nuclear ribonucleoprotein E2, hnRNP E2
Select a Size
$182.00
Select a Size
About This Item
$182.00
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
PC 81-1, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... PCBP2(5094)
General description
Specificity
Immunogen
Application
Inflammation & Immunology
Immunocytochemistry Analysis: A representative lot detected PCBP2 (hnRNP E2) in Immunocytochemistry applications (Fitzgerald, K.D., et. at. (2011). PLoS Pathog. 7(7):e1002127).
Western Blotting Analysis: 1 µg/mL from a representative lot detected PCBP2 (hnRNP E2) in SK-N-SH cell lysate.
Quality
Western Blotting Analysis: 0.1 µg/mL of this antibody detected PCBP2 (hnRNP E2) in HeLa cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service