MABF2090
Anti-Ly49A Antibody, clone A1
clone A1, from mouse
Synonym(s):
T-cell surface glycoprotein YE1/48, Lymphocyte antigen 49a, Ly-49a, T lymphocyte antigen A1
Select a Size
About This Item
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
A1, monoclonal
species reactivity
mouse
packaging
antibody small pack of 25 μg
technique(s)
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2aκ
UniProt accession no.
target post-translational modification
unmodified
Gene Information
mouse ... Klra1(16627)
General description
Specificity
Immunogen
Application
Immunocytochemistry Analysis: A representative lot detected Ly49A in Immunocytochemistry applications (Nagasawa, R., et. al. (1987). J Immunol. 138(3):815-24; Back, J., et. al. (2007). Proc Natl Acad Sci U S A. 104(10):3978-83).
Flow Cytometry Analysis: A representative lot detected Ly49A in Flow Cytometry applications (Nagasawa, R., et. al. (1987). J Immunol. 138(3):815-24; Yokoyama, W.M., et. al. (1989). J Immunol. 143(4):1379-86; Back, J., et. al. (2007). Proc Natl Acad Sci U S A. 104(10):3978-83; Silver, E.T., et. al. (2000). J Immunol. 165(4):1771-81).
Quality
Flow Cytometry Analysis: 1 µg of this antibody detected Ly49A in 1X10E6 EL-4 T lymphoma cells.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service