Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2031M

Sigma-Aldrich

Anti-CD1b Antibody, clone BCD1b3.1

clone BCD1b3.1, from mouse

Synonym(s):

T-cell surface glycoprotein CD1b

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$182.00
100 μG
$401.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$182.00
100 μG
$401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$182.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BCD1b3.1, monoclonal

species reactivity

guinea pig, human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
neutralization: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD1B(910)

General description

T-cell surface glycoprotein CD1b (UniProt: P29016; also known as CD1b) is encoded by the CD1B gene (Gene ID: 910) in human. CD1b is an antigen-presenting protein that binds self and non-self lipid and glycolipid antigens and presents them to T-cell receptors on natural killer T-cells. The most prominent known function of CD1b molecules is to associate with lipids from pathogens in order to stimulate T cells and induce host defense mechanisms. CD1b is expressed on cortical thymocytes, certain T-cell leukemia, and other tissues. Two isoforms of CD1b are reported that are produced by alternative splicing. CD1b has an extracellular domain (aa 18-3-3), a helical domain (aa 304-324), and a cytoplasmic tail (aa 329-332). The newly synthesized CD1b molecules follow an intracellular transport pathway that leads it to the cell surface from endoplasmic reticulum and subsequently to internalization from the plasma membrane into the endocytic pathway. After internalization, CD1b molecules are sorted for transport to lysosomes in the sorting endosome. (Ref.: Briken, V., et al. (2002). EMBO J. 21(4); 825-834).

Specificity

Clone BCD1b3.1 detects T-cell surface glycoprotein CD1b in human and guinea pig.

Immunogen

Isolated human CD1+ monocytes.

Application

Anti-CD1b, clone BCD1b3.1, Cat. No. MABF2031, is a mouse monoclonal antibody that detects T-cell surface glycoprotein CD1b and it has been tested for use in Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition assays, and Neutralizing function.
Immunohistochemistry Analysis: A representative lot detected CD1b in Immunohistochemistry applications (Dascher, C.C., et. al. (1999). J immunol. 163(10):5478-88).

Flow Cytometry Analysis: A representative lot detected CD1b in Flow Cytometry applications (Hiromatsu, K., et. al. (2002). Immunology. 106(2):159-72; Torrelles, J.B., et. al. (2011). J Biol Chem. 286(41):35438-46; Dascher, C.C., et. al. (1999). J immunol. 163(10):5478-88; Sieling, P.A., et. al. (2005). J Immunol. 174(5):2637-44; Facciotti, F., et. al. (2011). Proc Natl Acad Sci USA. 108(34):14228-33).

Immunoprecipitation Analysis: A representative lot detected CD1b in Immunoprecipitation applications (Briken, V., et. al. (2002). EMBO J. 21(4):825-34; Dascher, C.C., et. al. (1999). J immunol. 163(10):5478-88).

Neutralizing Analysis: A representative lot detected CD1b in Neutralizing applications (Torrelles, J.B., et. al. (2011). J Biol Chem. 286(41):35438-46).

Inhibition Analysis: A representative lot detected CD1b in Inhibition applications (Behar, S.M., et. al. (1995). J Exp Med. 182(6):2007-18).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in MOLT4 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CD1b in one million MOLT4 cells.

Target description

36.94 kda calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service