Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1973M

Sigma-Aldrich

Anti-Etanercept Antibody, clone ETA63C8

clone ETA63C8, from mouse

Synonym(s):

Enbrel

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$182.00
200 μL
$401.00

$182.00


Check Cart for Availability


Select a Size

Change View
25 μL
$182.00
200 μL
$401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$182.00


Check Cart for Availability

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

ETA63C8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
neutralization: suitable

isotype

IgG1κ

target post-translational modification

unmodified

Related Categories

General description

Etanercept is a chimeric fusion protein consisting of the extracellular ligand-binding portion of the p75 TNF receptor linked to the Fc portion of human IgG1. This Fc component contains the hinge, CH2 and CH3 regions, but not the CH1 region of IgG1. Etanercept binds to TNF-alpha and blocks its interactions with receptors. Hence, it is considered a TNF inhibitor that functions as a decoy receptor that binds to TNF. Although it mimics the inhibitory effects of naturally occurring soluble TNF receptors, the difference is that Etanercept has a greatly extended half-life in the bloodstream. Its speci fic activity is reported to be about 1.7 x 106 units/mg. Etanercept is shown to modulate biological responses produced by TNF, including expression of adhesion molecules responsible for leukocyte migration, serum levels of cytokines, and serum levels of matrix metalloproteinase-3. It is also shown to affect animal models of inflammation, including murine collagen-induced arthritis. Clone ETA63C8 can bind to Etanercept and block its activity.

Specificity

Clone ETA63C8 detects Etanercept in human samples. It can bind to Etanercept and block its activity.

Immunogen

Chimeric fusion protein cotaining the extracellular ligand-binding portion of the p75 TNF receptor linked to the Fc portion of human IgG1.

Application

Anti-Etanercept, clone ETA63C8, Cat. No. MABF1973, is a mouse monoclonal antibody that can bind to Etanercept and has been tested for use in ELISA and Neutralizing.
Neutralizing Analysis: A representative lot neutralized Etanercept in Neutralizing applications (Detrez, I., et. al. (2016). J Immunol. 196(6):2879-84).

ELISA Analysis: A representative lot detected Etanercept in ELISA applications (Detrez, I., et. al. (2016). J Immunol. 196(6):2879-84).

ELISA Analysis: 4 µg/mL from a representative lot detected Etanercept in human serum with AS (ankylosing spondylitis) (Courtesy of Professor Ann Gils, Ph.D., KU Leuven Belgium).
Research Category
Inflammation & Immunology

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1.

Target description

150 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Buongiorno, il prodotto MABF1973M (Anti-Etanercept Antibody, clone ETA63C8) presente due formati, 25 uL e 200 uL, ma che concentrazione sono? Grazie

    1 answer
    1. The concentration of this antibody is 0.5 mg/mL. This information is reported in the lot-specific Certificate of Analysis. See the link below to access this document:
      https://www.emdmillipore.com/product/Anti-Etanercept-Antibody-clone-ETA63C8,MM_NF-MABF1973#anchor_COA

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service