Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1826

Sigma-Aldrich

Mouse IgG2b Negative Control, clone MPC-11, redFluor® 710 conjugate Antibody

clone MPC-11, from mouse, purified by affinity chromatography

Synonym(s):

Immunoglobulin G2b, IgG2b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$208.00

$208.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$208.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$208.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

clone

MPC-11, monoclonal

purified by

affinity chromatography

technique(s)

flow cytometry: suitable

isotype

IgG2b

target post-translational modification

unmodified

General description

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This mouse monoclonal Mouse IgG2b Negative Control, clone MPC-11, redFluor® 710 conjugate, Cat. No. MABF1826 is validated for use in Flow Cytometry.

Physical form

Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

redFluor is a registered trademark of Tonbo Biotechnologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service