Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1823

Sigma-Aldrich

Rat IgG2a Negative Control, clone 2A3, redFluor® 710 conjugate Antibody

clone 2A3, from rat, purified by affinity chromatography

Synonym(s):

Immunoglobulin G2a, IgG2a

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$188.00

$188.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$188.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$188.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

clone

2A3, monoclonal

purified by

affinity chromatography

technique(s)

flow cytometry: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

General description

The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Application

This rat monoclonal Rat IgG2a Negative Control, clone 2A3, redFluor® 710 conjugate, Cat. No. MABF1823 is validated for use in Flow Cytometry.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

redFluor is a registered trademark of Tonbo Biotechnologies

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service