MABF148B
Anti-Ly-6G (mouse), violetFluor® 450, clone RB6-8C5 Antibody
clone RB6-8C5, from rat, violetFluor® 450
Synonym(s):
Lymphocyte antigen 6G (Ly-6G), Ly-6G.1, Ly-6G
Select a Size
About This Item
biological source
rat
Quality Level
conjugate
violetFluor® 450
antibody form
purified antibody
antibody product type
primary antibodies
clone
RB6-8C5, monoclonal
species reactivity
mouse
packaging
antibody small pack of 25 μg
technique(s)
flow cytometry: suitable
isotype
IgG2bκ
UniProt accession no.
target post-translational modification
unmodified
Gene Information
mouse ... Ly6G(546644)
Related Categories
General description
Immunogen
Application
Inflammation & Immunology
Immunoglobulins & Immunology
Quality
Flow Cytometry Analysis: 0.06 μg from a representative lot detected Ly-6G in one million C57Bl/6 bone marrow cells.
Physical form
Storage and Stability
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service