Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF1070

Sigma-Aldrich

Anti-C-Reactive Protein Antibody, clone 2A8.1

clone 2A8.1, from mouse

Synonym(s):

C-reactive protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CRP(1401)

General description

C-reactive protein (UniProt P02741) is encoded by the CRP (also known as PTX1) gene (Gene ID 1401) in human. CRP is the classical acute phase reactant produced by the liver and its circulating concentration increases rapidly in a cytokine-mediated response to tissue injury, infection, and inflammation. It is an annular, pentameric protein that binds to lysophosphatidylcholine expressed on the surface of dead or dying cells and bacteria in order to activate the complement system via the C1Q complex. Interleukin-6 (IL-6), produced by macrophages in response to injury and infection, is a major trigger for the synthesis of CRP by the liver. It plays a major role in early host defense by promoting agglutination, bacterial capsular swelling, phagocytosis, and complement fixation through its calcium-dependent binding to phosphorylcholine. It can also interact with DNA and histones and scavenge nuclear material released from damaged circulating cells.

Specificity

Clone 2A8.1 targets an epitope within the C-terminal half of human C-reactive protein.

Immunogen

GST-tagged recombinant human C-reactive protein C-terminal fragment.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected C-reactive protein in human cerebral cortex, kidney, and liver tissue sections.
This mouse monoclonal Anti-C-Reactive Protein, clone 2A8.1 Antibody, Cat. No. MABF1070 is validated for use in Western Blotting and Immunohistochemistry (Paraffin), for the detection of C-Reactive protein.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected C-reactive protein in 10 µg of human liver tissue lysate.

Target description

~25 kDa observed. 25.04/23.05 kDa (with/without signal peptide a.a. 1-18 sequence) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole E Behrens et al.
mBio, 10(2) (2019-03-14)
In patients with invasive fungal diseases, there is often little cellular inflammatory response. We tested the idea that binding of the human constitutive plasma protein serum amyloid P component (SAP) (also called PTX2) to Candida albicans dampens the innate immune

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service