Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE983

Sigma-Aldrich

Anti-Prohibitin Antibody, clone 2E12

clone 2E12, from mouse

Synonym(s):

Prohibitin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E12, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHB(5245)

General description

Prohibitin is an inhibitor of DNA synthesis that is widely expressed across various tissues. Prohibitin influences proliferation, and may also have a role in regulating mitochondrial respiration activity and in aging. Expression levels of prohibitin in fibroblasts have been found to decrease heterogeneously as cellular aging occurs.

Immunogen

Corresponding to human Prohibitin.

Application

Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected Prohibitin in 1X10E6 Jurkat T cells.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Prohibitin in MRC5 cells.
Flow Cytometry Analysis: A representative lot detected Prohibitin in Jurkat cells (Thomas Moran, Mount Sinai School of Medicine, New York, NY).
Immunocytochemistry Analysis: A representative lot detected Prohibitin in HeLa cells (Thomas Moran, Mount Sinai School of Medicine, New York, NY).
This Anti-Prohibitin Antibody, clone 2E12 is validated for use in Immunocytochemistry and Flow Cytometry and Immunoprecipitation for the detection of Prohibitin.

Quality

Evaluated by Immunocytochemistry in HEK293 and NIH/3T3 cells.

Immunocytochemistry Analysis: A 1:50 dilution of this antibody detected Prohibitin in HEK293 and NIH/3T3 cells.

Target description

30 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service