Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE966

Sigma-Aldrich

Anti-DNA polymerase II subunit A Antibody, clone 9F11.1

clone 9F11.1, from mouse

Synonym(s):

DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9F11.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

DNA polymerase epsilon catalytic subunit A (EC 2.7.7.7; UniProt Q07864; also known as DNA polymerase II subunit A) is encoded by the POLE (also known as CRCS12, FILS, POLE1) gene (Gene ID 5426) in human. DNA polymerase II is one of the six known mammalian template-directed DNA polymerases. It is composed of two subunits (261 kDa and 59 kDa). The larger catalytic subunit is a B-type polymerase family member, containing polymerase and intrinsic 3′-5′ exonuclease activities in its N-terminal part and a putative zinc-finger domain in its C-terminus. POLE gene mutations are linked to colorectal cancer, facial dysmorphism, immunodeficiency, livedo, and short stature.

Specificity

Clone 9F11.1 reacts with the 220 kD catalytic subunit of DNA polymerase epsilon. It does not recognize DNA polymerases alpha, beta or delta.

Immunogen

Epitope: Near N-terminus
GST-tagged recombinant protein corresponding to human DNA polymerase II subunit A near the N-terminus.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-DNA polymerase II subunit A Antibody, clone 9F11.1 is validated for use in Western Blotting for the detection of DNA polymerase II subunit A .

Quality

Evaluated by Western Blotting in human erythroleukemia K562 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected DNA polymerase II subunit A in 10 µg of human erythroleukemia K562 cell lysate.

Target description

~260 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine A M Weber et al.
Acta neuropathologica communications, 11(1), 184-184 (2023-11-22)
Pathogenic germline variants in the DNA polymerase genes POLE and POLD1 cause polymerase proofreading-associated polyposis, a dominantly inherited disorder with increased risk of colorectal carcinomas and other tumors. POLE/POLD1 variants may result in high somatic mutation and neoantigen loads that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service