Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE894

Sigma-Aldrich

Anti-Eri1/Exoribonuclease 1 Antibody, clone 5G8

clone 5G8, from rat

Synonym(s):

3′-5′ exoribonuclease 1, 3′-5′ exonuclease ERI1, Eri-1 homolog, Histone mRNA 3′-exonuclease 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G8, monoclonal

species reactivity

mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Eri1(67276)

General description

The protein named 3’-5’ exoribonuclease 1 or 3′-5′ exonuclease ERI1, Eri-1 homolog, Histone mRNA 3′-end-specific exoribonuclease, Histone mRNA 3′-exonuclease 1, or Protein 3′hExo (HEXO), and encoded by the human gene ERI1/ 3’EXO,/THEX1 is an RNA exonuclease that binds to the 3′-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. Eri1 also can degrade 3’ overhangs on short interfering RNAs (siRNAs) suggestion a possible wider role in RNA interference, and finally Eri1 also processes other RNAs, including ribosomal RNAs and Eri1 is part of a pre-mRNA processing complex and can bind ribosomal subunits and assembled ribosomes too. Recent research also has identified Eri1 as a regulator of micro RNA and plays a role in miRNA homeostasis, lymphocyte development and antiviral functions. Eri1 is localized to the cytoplasm and nucleus, as well as, the nucleolus in normal cells.

Immunogen

Recombinant protein corresponding to mouse Eri1/Exoribonuclease 1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
This Anti-Eri1/Exoribonuclease 1 Antibody, clone 5G8 is validated for use in western blotting & IP for the detection of Eri1/Exoribonuclease 1.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Eri1/Exoribonuclease 1 in 10 µg of mouse testis tissue lysate.

Western Blotting Analysis: A representative lot from an independent laboratory detected Eri1/Exoribonuclease 1 in MEF cell lysate (Hoefig, K. P., et al. (2012). Nat Struct Mol Biol. 20(1):73-81.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Eri1/Exoribonuclease 1 from MEF cell lysate (Hoefig, K. P., et al. (2012). Nat Struct Mol Biol. 20(1):73-81.).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Eri1/Exoribonuclease 1 in 10 µg of NIH/3T3 cell lysate.

Target description

~40 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Declercq et al.
Nucleic acids research, 48(18), 10428-10440 (2020-09-23)
Cellular exonucleases involved in the processes that regulate RNA stability and quality control have been shown to restrict or to promote the multiplication cycle of numerous RNA viruses. Influenza A viruses are major human pathogens that are responsible for seasonal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service