Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE882

Sigma-Aldrich

Anti-B-cell lymphoma/leukemia 11B, clone 3A8.2 Antibody

clone 3A8.2, from mouse

Synonym(s):

B-cell lymphoma/leukemia 11B, BCL-11B, B-cell CLL/lymphoma 11B, COUP-TF-interacting protein 2, Radiation-induced tumor suppressor gene 1 protein, hRit1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A8.2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCL11B(64919)

General description

B-cell lymphoma/leukemia 11B, also known as B-cell lymphoma/leukemia 11B (BCL-11B), B-cell CLL/lymphoma 11B, COUP-TF-interacting protein 2, Radiation-induced tumor suppressor gene 1 protein, hRit1, and encoded by the gene name BCL11B/ CTIP2/ RIT1, is a tumor-suppressor protein involved in t-cell lymphomas. B-cell lymphoma/leukemia 11B have been shown to play a role in the p53-signaling pathway. B-cell lymphoma/leukemia 11B may be a key regulator of both differentiation and survival during thymocyte development. B-cell lymphoma/leukemia 11B represses transcription through direct, tfcoup2-independent binding to a gc-rich response element. B-cell lymphoma/leukemia 11B interacts with tfcoup1, sirt1, arp1 and ear2. B-cell lymphoma/leukemia 11B is highly expressed in brain and in malignant T-cell lines derived from patients with adult T-cell leukemia/lymphoma.

Immunogen

GST-tagged recombinant protein corresponding to human B-cell lymphoma/leukemia 11B.

Application

Anti-B-cell lymphoma/leukemia 11B, clone 3A8.2 is an antibody against B-cell lymphoma/leukemia 11B for use in WB, ICC.
Immunocytochemistry Analysis: 20 µg/mL from a representative lot detected B-cell lymphoma/leukemia 11B in Jurkat cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected B-cell lymphoma/leukemia 11B in 10 µg of Jurkat cell lysate.

Target description

~94 and ~86 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service