Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE643

Sigma-Aldrich

Anti-monomethyl Histone H3 (Lys79), clone RM147 Antibody

clone RM147, from rabbit

Synonym(s):

Histone H3.3, Histone H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$405.00

$405.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$405.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$405.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM147, monoclonal

species reactivity

human

technique(s)

multiplexing: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys79)

Gene Information

human ... H3F3B(3021)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. Lysine methylation occurs in three distinct states, having either one (me1), two (me2) or three (me3) methyl groups attached to the amine group of the lysine side chain. Methylation of Lysine 79 in Histone 3 acts as a marker of inactive chromatin regions. Methylation by Dot1 methylase on lysine-79 (Lys79) of Histone H3 is criticial for transcriptional silencing, and it is thought that silencing proteins such as Sir3 function by blocking Dot1 methylation.

Specificity

This antibody reacts to Histone H3 monomethylated at Ly79 (K79me1). No cross reactivity with dimethylated Lys79 (K79me2) or trimethylated
Lys79 (K79me3), or other methylations in histone H3.

Immunogen

A linear peptide corresponding to human Histone H3 monomethylated at Lys79.

Application

Multiplex Analysis (Mplex): 0.1-0.5 μg/mL from a representative lot was used in Multiplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
This Anti-monomethyl Histone H3 (Lys79), clone RM147 Antibody is validated for use in Western Blotting, Multiplexing for the detection of monomethyl Histone H3.

Quality

Evaluated by Western Blotting in HeLa acid extract and HeLa cell lysate.

Western Blotting Analysis (WB): 0.5 μg/mL of this antibody detected monomethyl Histone H3 (Lys79) in HeLa cell lysate.

Target description

~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service