Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE596

Sigma-Aldrich

Anti-HMGN2, clone EPR7091, Rabbit Monoclonal Antibody

clone EPR7091, from rabbit

Synonym(s):

Non-histone chromosomal protein HMG-17, High mobility group nucleosome-binding domain-containing protein 2, HMGN2, HMG17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR7091, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HMGN2(3151)

General description

HMGN2 (High Mobility Group Nucleosomal binding domain 2), also known as HMG17, is an abundant, nonhistone chromosomal protein that binds preferentially to nucleosomal core particles of mammalian chromatin. HMGN2 binds to the inner side of the nucleosomal DNA thus altering the interaction between the DNA and the histone octamer. It acts as a chromatin-specific transcriptional coactivator that increases the efficiency of initiation of transcription by RNA polymerase II. HMGN2 also plays a role in chromatin assembly and conformation.

Immunogen

Synthetic peptide corresponding to human HMGN2.

Application

Immunohistochemistry Analysis: A 1:250-500 dilution from a representative lot detected HMGN2 in human colon tissue.

Produced in collaboration with Epitomics
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-HMGN2 antibody is validated for use in WB, IH for the detection of HMGN2.

Quality

Evaluated by Western Blotting in HL60, K562, HeLa, SH-SY5Y, and Jurkat cell lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected HMGN2 in HL60, K562, HeLa, SH-SY5Y, and Jurkat cell lysates.

Target description

~19 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service