Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE571

Sigma-Aldrich

Anti-MPS-1, clone 4A12 Antibody

ascites fluid, clone 4A12, from mouse

Synonym(s):

40S ribosomal protein S27, Metallopan-stimulin 1, MPS-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4A12, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RPS27(6232)

General description

40S ribosomal protein S27, also known as Metallopan-stimulin 1 (MPS-1), and encoded by the gene name MPS1 or RPS27, belongs to the ribosomal protein S27e family. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. MPS-1 is a component of the 40S subunit and contains a C4-type zinc finger domain that can bind to zinc. MPS-1 is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that MPS-1 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3.

Immunogen

Purified recombinant fragment of human MPS-1 expressed in E. Coli

Application

Anti-MPS-1, clone 4A12 Antibody detects level of MPS-1 & has been published & validated for use in MPS-1.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected MPS-1 in human lung cancer and brain tissues.

Target description

9 kDa calculated

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangling Chen et al.
Theranostics, 8(9), 2348-2360 (2018-05-04)
The X-linked deubiquitinase, USP9X, is implicated in multiple cancers by targeting various substrates. Increased expression of USP9X is observed in non-small-cell lung cancer (NSCLC) and is correlated with poor prognosis. However, the molecular mechanism for USP9X regulation of tumor cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service