MABE451
Anti-p31comet Antibody, clone E29.19.14
clone E29.19.14, from mouse
Synonym(s):
MAD2L1-binding protein, Caught by MAD2 protein
Select a Size
$472.00
Select a Size
About This Item
$472.00
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
E29.19.14, monoclonal
species reactivity
human
technique(s)
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... MAD2L1BP(9587)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Chromatin Biology
Immunoprecipitation Analysis: A representative lot from an independent laboratory detected p31comet in HeLa cell lysates, which were untreated (asynchronous), arrested in nocodazole for 16 hr, or released in fresh medium for 2 hr (Varetti, G., et al. (2011). 44(5):710-720.).
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected p31comet in 10 µg of HeLa cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service