Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE435

Sigma-Aldrich

Anti-phospho Histone H3 (Thr11) Antibody, clone N123/48

clone N123/48, 0.5 mg/mL, from mouse

Synonym(s):

Histone H3.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N123/48, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

concentration

0.5 mg/mL

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr11)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)
rat ... H3C1(679994)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone proteins are highly post-translationally modified, and Histone H3 is the most extensively modified of the five histones. Histone H3 sequence variants and variable modification states are thought to play a role in the dynamic and long term regulation of genes. Di- and tri-methylation of histone H3 are gene silencing marks, while removal of these marks by the demethylase genes Jmjd1a and Jmjd2c leads to gene activation.

Immunogen

Linear peptide corresponding to rat Histone H3 phosphorylated at Thr11.

Application

Immmunocytochemistry Analysis: A 1:50 dilution from a representative lot detected phospho Histone H3 (Thr11) in HeLa cells.

Dot Blot Analysis: A 1:5,000 dilution from a representative lot detected phospho Histone H3 (Thr11) in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).

Dylight is a registered trademark of Thermo Fisher Scientific.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H3 (Thr11) Antibody, clone N123/48 is validated for use in western blotting, ICC & dot blot for the detection of phospho Histone H3 (Thr11).

Quality

Evaluated by Western Blotting in Colcemid treated and untreated HeLa acid extract.

Western Blotting Anlaysis: 1:5,000 dilution of this antibody detected phospho Histone H3 (Thr11) in Colcemid treated HeLa acid extract and not in untreated HeLa acid extract.

Target description

~17 kDa observed

Linkage

Replaces: MABE468

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service