Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE431

Sigma-Aldrich

Anti-PRMT1 Antibody, clone 7D2

clone 7D2, from rat

Synonym(s):

Protein arginine N-methyltransferase 1, Histone-arginine N-methyltransferase PRMT1, Interferon receptor 1-bound protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D2, monoclonal

species reactivity

rat, mouse, human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... PRMT1(3276)

General description

The methylation of arginine is mediated by the Protein Arginine Methyl Transferase (PRMT) family of enzymes. These enzymes transfer the methyl group from S-adenosyl-Lmethionine to the guanidino nitrogen atoms of an arginine residue. Arginine methylation was found to be an important modification in signal transduction, transcription, RNA transport, and splicing. PRMT1, also known as HRMT1L2 and IR1B4, was isolated through its interaction with BTG1 and TIS21, proteins that are important in cell quiescence. PRMT1 is a 361 amino acid protein; its splicing variants differ at the N-terminus, giving rise to three isoforms of 343, 361, and 347 amino acids (variants 1, 2, and 3 respectively). Proteins involved in the metabolism of RNA are substrates for PRMT1, among them Sam68.

Immunogen

BSA-conjugated linear peptide corresponding to human PRMT1.

Application

Detect PRMT1 using this rat monoclonal antibody, Anti-PRMT1 Antibody & clone 7D2 validated for use in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Western Blotting Analysis: A representative lot from an independent laboratory detected PRMT1 in E. Coli containing untagged PRMT1, E.Coli extract containing His-tagged PRMT1, E.Coli extract containing GST- tagged PRMT1, and in DG75 whole cell extract (Gross, H., et al. (2012). PLoS ONE. 7(8):e42106.).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PRMT1 in 10 µg of HEK293 cell lysate.

Target description

~ 42 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service