Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE354

Sigma-Aldrich

Anti-Uracil-DNA glycosylase Antibody, clone 8G10.1

clone 8G10.1, from mouse

Synonym(s):

UDG, Uracil-DNA glycosylase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$365.00

$365.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$365.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$365.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G10.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UNG(7374)

General description

Uracil-DNA glycosylase (EC 3.2.2.27; UniProt P13051; also known as UDG) is encoded by the UNG (also known as DGU, HIGM4, HIGM5, UNG1, UNG2, UNG15) gene (Gene ID 7374) in human. Uracil-DNA glycosylases catalyze the release of uracil from DNA molecules by cleaving the N-glycosylic bond and initiating the base-excision repair (BER) pathway that plays an important role in mutagenesis prevention. Uracil bases in DNA can result from cytosine deamination or misincorporation of dUMP instead of dTTP during DNA synthesis. Alternative promoter usage and splicing of UNG gene lead to two different isoforms, the mitochondrial UNG1 and the nuclear UNG2. The mammalian shelterin complex contains the six proteins (TRF1, TRF2, RAP1, TIN2, POT1, and TPP1), the complex occupies the end of the telomeres and protect telomeres against inappropriate DNA repair when telomeres are not being lengthened. UNG deficiency in primary mouse hematopoietic cells is shown to affect POT1 (Protector of Telomeres 1) recognition of the telomere repeats due to increased uracil incorporation in telomeres TTAGGG repeats, which in turn leads to abnormal telomere lengthening.

Specificity

Expected to react with both spliced isoforms (UNG1 & UNG2) of human Uracil-DNA glycosylase.

Immunogen

Epitope: Near C-terminus
Linear peptide corresponding to human Uracil-DNA glycosylase near the C-terminus.

Application

Anti-Uracil-DNA glycosylase Antibody, clone 8G10.1, Cat. No. MABE354, is a highly specific mouse monoclonal antibody, that targets UNG and has been tested in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Uracil-DNA glycosylase in human skeletal muscle and human cardiac tissues.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Uracil-DNA glycosylase in 10 µg of HepG2 cell lysate.

Target description

~34 kDa observed (33.9 kDa/UNG1 & 34.6 kDa/UNG2 calculated). Immunogen sequence should allow to detect both isoform 1 and 2. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service