MABE315
Anti-JMJD2C Antibody, clone 6A3.1
clone 6A3.1, 0.5 mg/mL, from mouse
Synonym(s):
Lysine-specific demethylase 4C, Gene amplified in squamous cell carcinoma 1 protein, GASC-1 protein, JmjC domain-containing histone demethylation protein 3C, Jumonji domain-containing protein 2C
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6A3.1, monoclonal
species reactivity
human
concentration
0.5 mg/mL
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... KDM4C(23081)
General description
Immunogen
Application
Alexa Fluor™ is a registered trademark of Life Technologies.
Epigenetics & Nuclear Function
Chromatin Biology
Quality
Western Blotting Analysis: 1 µg/mL of this antibody detected JMJD2C in 10 µg of HeLa neclear extract.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service