Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABE1118

Sigma-Aldrich

Anti-BRG1/BAF190A Antibody, clone 5B7

clone 5B7, from rat

Synonym(s):

Transcription activator BRG1, ATP-dependent helicase SMARCA4, BAF190A, BRG1-associated factor 190A, hSNF2b, Mitotic growth and transcription activator, Protein brahma homolog 1, Protein BRG-1, SNF2-beta, SWI/SNF-related matrix-associated actin-dependent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified antibody

antibody product type

primary antibodies

clone

5B7, monoclonal

species reactivity

mouse, human, monkey

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMARCA4(6597)

General description

Transcription activator BRG1 (UniProt P51532; also known as ATP-dependent helicase SMARCA4, BAF190A, BRG1-associated factor 190A, hSNF2b, Mitotic growth and transcription activator, Protein brahma homolog 1, Protein BRG-1, SNF2-beta, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4) is encoded by the SMARCA4 (also known as BAF190A, BRG1, MRD16, RTPS2, SNF2B, SNF2L4) gene (Gene ID 6597) in human. The SWI/SNF-like BRG1/BRM-associated factor (BAF) chromatin-remodeling complexes drive conformational changes of nucleosomes in an ATP-dependent manner and play important roles in cell-cycle progression, DNA repair, and development. The SWI/SNF complexes contain 10-12 interchangeable subunits, with either BRG1/SMARCA4 or BRM/SMARCA2 as the ATPase subunit. Brg1 is also shown to maintain Polycomb-mediated repression of non-mesodermal developmental regulators. Brg1-knockout leads to disruption of murine ESC cardiomyocyte differentiation and dysregulation of lineage-specific gene expression during mesoderm induction.

Specificity

Expected to react with all five spliced human isoforms and both murine spliced isoforms reported by UniProt (P51532 & Q3TKT4).

Immunogen

Epitope: N-terminal region.
GST-tagged recombinant internal fragment from the N-terminal region of human BRG1/BAF190A.

Application

Anti-BRG1 Antibody, clone 5B7 is an antibody against BRG1/BAF190A for use in Western Blotting, Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected BRG1/BAF190A in HeLa cell lysate.
Western Blotting Analysis: A representative lot detected BRG1/BAF190A in human HeLa, monkey Cos-1, and murine L929 cell lysates (Courtesy of Dr. Taro Tachibana, Osaka City University, Japan).
Immunocytochemistry Analysis: A representative lot detected BRG1/BAF190A in HeLa cells (Courtesy of Dr. Taro Tachibana, Osaka City University, Japan).
Immunocytochemistry Analysis: Clone 5B7 hybridoma culture supernatant detected BRIG1 co-localization with microphthalmia transcription factor (Mitf) nuclear immunoreactivity by fluorescent immunocytochemistry staining of 4% paraformaldehyde-fixed, 0.5% Triton X-100-permeabilized, NIH/3T3-derived B22 cells that harbors a tetracycline-inducible dominant-negative (DN) human BRG1 K798R construct regardless of DN BRG1 K798R induction by tetracycline treatment (de la Serna, I.L., et al. (2006). J. Biol. Chem. 281(29):20233-20241).

Quality

Evaluated by Western Blotting in murine L929 fibroblast lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected BRG1/BAF190A in murine L929 fibroblast lysate.

Target description

~185 kDa observed. 184.6 kDa (human isoform 1), 181.3 kDa (human isoform 2), 181.6 kDa (human isoform 3), 181.7 kDa (human isoform 4), 181.3 kDa (human isoform 5), 181.4 kDa (mouse isoform 1), and 181.5 kDa (mouse isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service