Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE110

Sigma-Aldrich

Anti-FOXA2 Antibody, clone 3C10

clone 3C10, from mouse

Synonym(s):

Hepatocyte nuclear factor 3-beta, HNF-3-beta, HNF-3B, Forkhead box protein A2, Transcription factor 3B, TCF-3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C10, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXA2(3170)

General description

Also known as hepatocyle nuclear factor 3-beta (HNF-3B), Forkhead box protein A2 (FOXA2) belongs to the forkhead class of DNA-binding proteins. This family of transcription factors influences gene expression in endodermally-derived tissues such as lung, liver, stomach, pancreas, and intestine. Foxa2 has also been detected in ectodermally-derived tissues such as kidney, uterus and vagina, and seminal and coagulating glands during development. Several studies show that FOXA proteins help facilitiate the binding of several nuclear receptors to their respective targets in a context-dependent manner, and are thereby implicated in various functions including cellular maintenance, differentiation, metabolism, and organogenesis. Aberrant function of FOXA proteins are attributed to diseases states such as diabetes and Parkinson′s disease.

Immunogen

Recombinant protein corresponding to the human FOXA2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-FOXA2 Antibody, clone 3C10 (Mouse Monoclonal Antibody) validated in WB to detect FOXA2 also known as Hepatocyte nuclear factor 3-beta, HNF-3-beta, HNF-3B.

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 1:500 dilution of this antibody detected FOXA2 on 10 µg of HepG2 cell lysate.

Target description

~50 kDa observed

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service