Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE1106

Sigma-Aldrich

Anti-WAPL Antibody, clone 1C7-B1

clone 1C7-B1, from mouse

Synonym(s):

WAPL, Wings apart-like protein homolog, Friend of EBNA2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C7-B1, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WAPAL(23063)

General description

Wings apart-like protein homolog (also known as Friend of Epstein-Barr virus nuclear protein 2) is a nuclear protein encoded by the WAPL (WAPAL) gene in human. Wapl functions as a negative regulator of cohesin–chromosome association and facilitates the progression of mitosis both during interphase and prophase. In interphase, Wapl destabilizes cohesin–chromosome interaction and thereby reduces the extent of cohesin-mediated chromosome looping. In prophase, mitotic kinases phosphorylate cohesin and its regulators. Wapl then triggers the release of phosphorylated cohesin from sister chromatids, allowing sister chromosome decatenation by topoisomerase II (Topo II). (PMID 24156811).

Immunogen

Recombinant protein corresponding to human WAPL.

Application

Anti-WAPL Antibody, clone 1C7-B1 is an antibody against WAPL for use in Western Blotting, Immunoprecipitation.
Immunoprecipitation Analysis: 1.0 mL tissue culture supernantant of this antibody immunoprecipitated WAPL in HeLa cell lysate (Professor Haruhiko Siomi, Keio University School of Medicine).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected WAPL in 10 µg of HeLa cell lysate.

Target description

~175 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service