Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE1093

Sigma-Aldrich

Anti-5-Hydroxymethylcytosine (5hmC) Antibody, clone EDL HMC 1A

clone EDL HMC 1A, from mouse

Synonym(s):

5hmC, 5-hmC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

EDL HMC 1A, monoclonal

species reactivity (predicted by homology)

all

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable

isotype

IgG2bκ

shipped in

ambient

target post-translational modification

unmodified

General description

5-Hydroxymethylcytosine (also known as 5-hmC) is the first oxidative product in the active demethylation of 5-methylcytosine (5mC). 5hmC is a DNA pyrimidine nitrogen base and is a predominantly stable DNA modification. It is produced by the action of three Ten-eleven translocation 1 (TET1) enzyme on 5mC. Each mammalian cell contains 5hmC, but its levels vary significantly depending on the cell type. Higher levels of 5hmC are found in neuronal cells and its level is shown to increase with age in hippocampus and cerebellum. It is also prevalent in embryonic stems cells and its reduced levels are shown to result in impaired self-renewal of stem cells. In the brain and in embryonic stem cells, 5hmC in enriched in promoters, gene bodies, and intergenic areas near genes and positively correlates with gene expression at these loci. 5hmC appears to play a role to promote gene expression during active demethylation. Significantly lower of 5hmC have been reported in cancer cell lines, melanoma, adenomas, and carcinomas compared to the normal surrounding tissue.

Specificity

Clone EDL HMC 1A targets 5-hydroxymethylcytosine (5hmC), but not 5-methylcytosine (5mC), 5-carboxylcytosine (5caC), 5-formylcytosine (5fC), or unmodified cytosine (C).
Target modification is not species-specific

Immunogen

BSA-conjugated 5-hydroxymethylcytidine.

Application

Anti-5-Hydroxymethylcytosine (5hmC), clone EDL HMC 1A, Cat. No. MABE1093, is a highly specific mouse monoclonal antibody that targets DNA with 5-hydroxymethylcytosine (5hmC) modification and has been tested in Dot Blot and Immunoprecipitation.
Dot Blot Analysis: 0.5 µg/mL from a representative lot detected 0.39-100 ng of DNA with 5-hydroxymethylcytosine (5hmC) modification, but not DNA with only 5-methylcytosine (5mC), 5-formylcytosine (5fC), 5-carboxylcytosine (5caC) modification or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).

Immunoprecipitation Analysis: 2 µg from a representative lot immunoprecipitated 5 ng of DNA with 5-hydroxymethylcytosine (5hmC) modification, but not DNA with only 5-methylcytosine (5mC), 5-formylcytosine (5fC), 5-carboxylcytosine (5caC) modification or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Dot Blot analysis of DNA cytosine modifications.

Dot Blot Analysis: 0.2 µg/mL of this antibody detected 50 ng of DNA with 5-hydroxymethylcytosine (5hmC), but not unmodified DNA or DNA with only 5-methylcytosine (5mC) modification.


Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Kazeminia et al.
Clinical science (London, England : 1979), 137(16), 1265-1283 (2023-08-22)
Scattered tubular-like cells (STCs) are differentiated renal tubular cells that during recovery from ischemic injury dedifferentiate to repair other injured renal cells. Renal artery stenosis (RAS), often associated with chronic inflammatory injury, compromises the integrity and function of STCs, but
Naba Farooqui et al.
Stem cells (Dayton, Ohio), 41(1), 50-63 (2022-10-18)
Atherosclerotic renal artery stenosis (ARAS) is associated with irreversible parenchymal renal disease and regenerative stem cell therapies may improve renal outcomes. Hypoxia preconditioning (HPC) may improve the regenerative functions of adipose tissue-derived mesenchymal stem cells (AMSC) by affecting DNA 5-hydroxymethylcytosine
Alfonso Eirin et al.
Cell death & disease, 15(6), 387-387 (2024-06-02)
Obesity exacerbates tissue degeneration and compromises the integrity and reparative potential of mesenchymal stem/stromal cells (MSCs), but the underlying mechanisms have not been sufficiently elucidated. Mitochondria modulate the viability, plasticity, proliferative capacity, and differentiation potential of MSCs. We hypothesized that
Kamalnath S Rajagopalan et al.
Cells, 11(11) (2022-06-11)
Scattered tubular-like cells (STCs) are dedifferentiated renal tubular cells endowed with progenitor-like characteristics to repair injured parenchymal cells. STCs may be damaged and rendered ineffective by renal artery stenosis (RAS), but the underlying processes remain unclear. We hypothesized that RAS
Alfonso Eirin et al.
American journal of physiology. Heart and circulatory physiology, 324(1), H14-H25 (2022-11-12)
Chronic kidney disease (CKD) is common in patients with heart failure and often results in left ventricular diastolic dysfunction (LVDD). However, the mechanisms responsible for cardiac damage in CKD-LVDD remain to be elucidated. Epigenetic alterations may impose long-lasting effects on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service