Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE1092

Sigma-Aldrich

Anti-5-Formylcytosine (5fC) Antibody, clone EDL FC-5

clone EDL FC-5, from mouse

Synonym(s):

5-Formylcytosine, 5fC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$472.00

$472.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$472.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

EDL FC-5, monoclonal

species reactivity (predicted by homology)

all

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable

isotype

IgG3λ

shipped in

wet ice

target post-translational modification

unmodified

General description

Four types of cytosine modifications have been identified in mammalian DNA, namely 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxylcytosine (5caC). While 5mC, 5hmC, and 5fC occur in embryonic stem (ES) cells and many cell types in all the major organs (e.g. brain, heart, lung and kidney), 5caC has only been detected in a few cell types at a 10 times lower level than 5fC. The sequential oxidation of 5mC to 5hmC, 5fC, and 5caC catalyzed by the ten-eleven translocation (TET) enzymes has been proposed as a potential route for the demethylation of 5mC back to cytosine (C). Thymine DNA glycosylase (TDG), for example, can excise 5fC and 5caC from DNA as part of the base excision repair (BER) pathway to restore the C-nucleotide. Emerging evidences suggest that the different modifications may be selectively recognized by various proteins, and methyl-CpG-binding protein 2 (MeCP2) binding to 5hmC has been proposed as a means of epigenetic regulation in neurons. The conversion of 5hmC to 5fC, in turn, induces an enhanced binding of DNA-repair-associated proteins to 5fC, which has been proposed as a signal for DNA demethylation.

Specificity

Clone EDL FC-5 specifically targets 5-formylcytosine (5fC), but not 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine (C).

Immunogen

BSA-conjugated 5-formylcytidine (5fC).
Epitope: 5-formylcytosine (5fC).

Application

Dot Blot Analysis: 0.5 µg/mL from a representative lot detected 1.25-100 ng of DNA with 5-formylcytosine (5fC) modification, but not DNA with only 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).
Immunoprecipitation Analysis: 2 µg from a representative lot immunoprecipitated 5 ng of DNA with 5-formylcytosine (5fC) modification, but not DNA with 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-5-Formylcytosine (5fC) Antibody, clone EDL FC-5 is validated for use in Dot Blot, Immunoprecipitation for the detection of 5-Formylcytosine (5fC).

Quality

Evaluated by Dot Blot analysis of DNA cytosine modifications.

Dot Blot Analysis: 2 µg/mL of this antibody detected 25-100 ng of DNA with 5-formylcytosine (5fC) modification, but not DNA with only 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine.

Target description

Variable, depending on the size(s) of the DNA molecule(s) with 5-formylcytosine (5fC) modification.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG3λ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman Thaler et al.
Nature communications, 13(1), 5883-5883 (2022-10-07)
Vitamin C deficiency disrupts the integrity of connective tissues including bone. For decades this function has been primarily attributed to Vitamin C as a cofactor for collagen maturation. Here, we demonstrate that Vitamin C epigenetically orchestrates osteogenic differentiation and function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service