Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1072

Sigma-Aldrich

Anti-CtIP Antibody, clone 11-1

clone 11-1, from mouse

Synonym(s):

CtBP-interacting protein, CtIP, Retinoblastoma-binding protein 8, RBBP-8, Retinoblastoma-interacting protein and myosin-like, RIM, Sporulation in the absence of SPO11 protein 2 homolog, SAE2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$429.00

$429.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$429.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

11-1, monoclonal

species reactivity

frog, mouse, human

technique(s)

affinity chromatography: suitable
immunodepletion: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RBBP8(5932)

General description

DNA endonuclease RBBP8 (UniProt: Q99708; also known CtBP-interacting protein, CtIP, Retinoblastoma-binding protein 8, RBBP-8, Retinoblastoma-interacting protein and myosin-like, RIM, Sporulation in the absence of SPO11 protein 2 homolog, SAE2) is encoded by the RBBP8 (also known as CTIP) gene (Gene ID: 5932) in human. RBBP-8 is an endonuclease that cooperates with the MRE11-RAD50-NBN (MRN) complex in processing meiotic and mitotic double-strand breaks (DSBs) by ensuring both resection and intrachromosomal association of the broken ends. RBBP8 is phosphorylated upon DNA damage (at Thr859) by ATR and this phosphorylated form binds to chromatin to initiate resection. The non-phosphorylated form does not bind to chromatin. It is reported that ATM activity is required for an early step in resection, leading to ATR activation, CtIP- phosphorylation, and accumulation of CtIP on chromatin. RBBP8 functions downstream of the MRN complex and promotes ATR activation and its recruitment to DSBs in the S/G2 phase facilitating the generation of ssDNA. It is localized predominantly in the nucleus and interacts with CTBP, with the C-terminal (BRCT) domains of BRCA1, and with the retinoblastoma protein. By interacting with BCRA1 it modulates transcriptional regulation, DNA repair, and cell cycle checkpoint control. Mutations in RBBP8 gene are known to cause Seckel syndrome 2, an autosomal recessive disorder characterized by proportionate dwarfism of prenatal onset associated with low birth weight, growth retardation, severe microcephaly, and mental retardation.

Specificity

Clone 11-1 specifically detects CtBP-interacting protein in human tonsils and pancreas.

Application

Anti-CtIP Antibody, clone 11-1, Cat. No. MABE1072, is a highly specific mouse monoclonal antibody that targets CtIP and has been used in Immunodepletion, Immunohistochemistry (Paraffin), Immunoaffinity Purification, and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected CtIP in normal human tonsil and human pancreatic cancer tissue.

Western Blotting Analysis: A representative lot detected CtIP in Western Blotting applications (Peterson, S.E., et. al. (2011). J Cell Biol. 194(5):705-20; Peterson, S.E., et. al. (2013). Mol Cell. 49(4):657-67).

Immunodepletion Analysis: A representative lot detected CtIP in Immunodepletion applications (Peterson, S.E., et. al. (2011). J Cell Biol. 194(5):705-20; Peterson, S.E., et. al. (2013). Mol Cell. 49(4):657-67).

Immunoaffinity Purification Analysis: A representative lot detected CtIP in Immunoaffinity Purification applications (Peterson, S.E., et. al. (2011). J Cell Biol. 194(5):705-20; Peterson, S.E., et. al. (2013). Mol Cell. 49(4):657-67).

Quality

Evaluated by Immunohistochemistry in human tonsil tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected CtIP in human tonsil tissue.

Target description

~120 kDa observed; 101.94 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service