Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABE1064M

Sigma-Aldrich

Anti-HsSAS-6 Antibody, clone 91.390.21

clone 91.390.21, from mouse

Synonym(s):

Spindle assembly abnormal protein 6 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

91.390.21, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SASS6(163786)

General description

Spindle assembly abnormal protein 6 homolog (UniProt: Q6UVJ0; also known as HsSAS-6) is encoded by the SASS6 (also known as SAS6) gene (Gene ID: 163786) in human. HsSAS-6 is a central scaffolding component of the centrioles that ensure their 9-fold symmetry. It functions to ensure that each centriole seeds the formation of a single procentriole per cell cycle. It is required for centrosome biogenesis and duplication and is essential for both for mother-centriole-dependent centriole duplication and deuterosome-dependent centriole amplification in multi-ciliated cells. It contains a 35 nM long coiled-coil domain that mediates homodimerization while the globular N-terminus links the dimers at an angle of 40 degrees to form the inner ring. Over-expression of SASS6 gene results in excess foci-bearing centriolar markers. Mutations in SASS6 are known to cause microcephaly 14 that is characterized by small head circumference, markedly reduced brain weight and disproportionately small cerebral cortex.

Specificity

Clone 91.390.21 detects HsSAS-6 protein in human cells. It targets an epitope with in 254 amino acids in the C-terminal half.

Immunogen

Epitope: C-terminus
His-tagged recombinant fragment corresponding to 254 amino acids from the C-terminal half of human Spindle assembly abnormal protein 6 homolog (HsSAS-6).

Application

Anti-HsSAS-6, clone 91.390.21, Cat. No. MABE1064, is a highly specific mouse monoclonal antibody that targets SASS6 and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 1 µg/mL from a representative lot detected HsSAS-6 in 10 µg of HeLa cell lysate.

Western Blotting Analysis: A representative lot detected HsSAS-6 in RNAi in U2OS cell lysates (Courtesy of Professor Dr. Erich Nigg, Ph.D; Biozentrum, University of Basel, Switzerland).

Immunofluorescence Analysis: A representative lot detected HsSAS-6 in RNAi followed by immunofluorescence (Courtesy of Professor Dr. Erich Nigg, PhD; Biozentrum, University of Basel, Switzerland).

Immunofluorescence Analysis: A representative lot detected HsSAS-6 in Immunofluorescence applications (Kleylein-Sohn, J. et. al. (2007). Dev Cell. 13(2):190-202; Sonnen, K..,et. al. (2012). Biol Open 1(10):965-76; Arquint, C., et al. (2012). J Cell Sci. 125(Pt 5):1342-52).

Immunoprecipitation Analysis: A representative lot detected HsSAS-6 in RNAi followed by immunoprecipitation (Courtesy of Professor Dr. Erich Nigg, PhD; Biozentrum, University of Basel, Switzerland).

Quality

Evaluated by Western Blotting in U20S cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected HsSAS-6 in 10 µg of U2OS cell lysate.

Target description

~74 kDa observed; 74.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service