Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE1033

Sigma-Aldrich

Anti-SMARCB1 Antibody, clone 2C2

clone 2C2, from mouse

Synonym(s):

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1, BRG1-associated factor 47, BAF47, Integrase interactor 1 protein, SNF5 homolog, hSNF5, SMARCB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SMARCB1(6598)

General description

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1 (SMARCB1), also known as BRG1-associated factor 47 (BAF47) or Integrase interactor 1 protein or SNF5 homolog (hSNF5), and encoded by the gene name SMARCB1 or BAF47, is part of a complex that relieves repressive chromatin structures, allowing the transcriptional machinery to access its targets more effectively. The encoded nuclear protein has also been shown to bind to and enhance the DNA joining activity of HIV-1 integrase. It has been found to be a tumor suppressor, and the loss of SMARCB1 has been associated with malignant rhabdoid tumors. Tumor cells of synovial sarcoma seem to have a post transcriptional regulatory mechanism for SMARCB1.

Immunogen

GST-tagged recombinant protein corresponding to human SMARCB1.

Application

Anti-SMARCB1 Antibody, clone 2C2 is an antibody against SMARCB1 for use in Western Blotting and Immunocytochemistry.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected SMARCB1 in 10 µg of HEK293 cell lysate.
Immunocytochemistry Analysis: A representative lot detected SMARCB1 in HeLa cells (Harada, A., et al. (2014). Mono. Antibodies in Immuno. and Immunotherapy. 33:49-51).
Western Blotting Analysis: A representative lot detected SMARCB1 in HeLa cells (Harada, A., et al. (2014). Mono. Antibodies in Immuno. and Immunotherapy. 33:49-51).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SMARCB1 in 10 µg of HeLa cell lysate.

Target description

~ 45 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service