Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABD170

Sigma-Aldrich

Anti-AFP Antibody, clone 6E6

clone 6E6, from mouse

Synonym(s):

Alpha-fetoprotein, FETA, Alpha-1-fetoprotein, Alpha-fetoglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

6E6, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AFP(174)

General description

Alpha fetoprotein (AFP) or alternatively known as Alpha-1-fetoprotein or Alpha-fetoglobulin is encoded by the human gene named AFP/HPAFP is an important early fetal blood protein. It is a secreted protein found in the plasma of fetuses older than 4 weeks of age and reaches the highest levels at 12th-16th week of gestation and then drops to trace amounts after birth. It is produced by the fetal liver and yolk sac normally. In normal adults the level is very low, but AFP can be a marker associated with various cancers and in particular with liver cancer and hepatomas. Its function during development is thought to be similar to that of serum albumin binding various ions and fatty acids as well as bilirubin, but while AFP is the most abundant protein found in the human fetus its complete function is still largely unknown. AFP as a biomarker in infants is associated with various diseases, however, because AFP levels change normally over a wide range in the developing fetus accurate aging is essential. High levels of AFP may suggest that the developing baby has a neural tube defect such as spina bifida or anencephaly. High levels of AFP may also suggest defects with the esophagus or a failure of baby’s abdomen to close. Low levels of AFP and abnormal levels of hCG and estriol may indicate that the developing baby has Trisomy 21 (Down syndrome), Trisomy 18 (Edwards Syndrome) or another type of chromosome abnormality. EMD-Millipore’s Anti-AFP mouse monoclonal antibody has been tested in western blot on HepG2 and SMMC-7721 cell lysates.

Immunogen

Purified recombinant fragment of human AFP expressed in E. Coli.

Application

Anti-AFP Antibody, clone 6E6 is a highly specific mouse monoclonal antibody & that targets α-Fetoprotein & has been tested in western blotting.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in HepG2 and SMMC-7721 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected AFP in HepG2 and SMMC-7721 cell lysates.

Optimal working dilutions must be determined by end user.

Target description

~69 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS containing up to 0.1% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
HepG2 and SMMC-7721 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service