Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC948

Sigma-Aldrich

Anti-CD1d/KRN7000 Antibody, clone L363

clone L363, from mouse

Synonym(s):

Antigen-presenting glycoprotein CD1d1, CD1d/KRN7000, CD1d.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L363, monoclonal

species reactivity

mouse

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd1D1(12479)

General description

Antigen-presenting glycoprotein CD1d1 (UniProt P11609; also known as CD1d.1, T-cell surface glycoprotein CD1d1) is encoded by the Cd1d1 (also known as Cd1.1, Ly-38, AI747460) gene (Gene ID 12479) in murine species. CD1d is a nonpolymorphic MHC class I-like molecule, which presents nonpeptide ligands (e.g. glycolipids) to Type 1 or invariant natural killer T (iNKT) cells. iNKT cells differ from conventional T cells in that they recognize glycolipid antigens presented by the nonclassical β2-microglobulin (β2m)-associated CD1d. Recognition of CD1d-presented glycolipids by iNKT cells elicits the rapid secretion of both T helper type 1- (TH1-) and T helper type 2- (TH2-) associated cytokines without the need for priming. Murine CD1d is initially produced with a signal peptide sequence (a.a. 1-21), the removal of which yields the mature CD1d with a large extracellular region (a.a. 22-305), a transmembrane domain (a.a. 306-326), and a short cytoplasmic tail (a.a. 327-336).

Specificity

Clone L363 selectively reacts with KRN7000-/α-GalCer-, OCH-, or C20:2- complexed mCD1d, but not glycolipid-free mCD1d. Clone L363 does not react with iGb3-complexed mCD1d, KRN7000-complexed hCD1b or KRN7000-complexed hCD1d (Yu, K.O., et al. (2007). J. Immunol. Methods. 323(1):11-23).

Immunogen

C-terminally BirA-His-tagged soluble mCD1d/mβ2m/KRN7000 complex cross-linked with purified protein derivative (PPD) from M. tuberculosis strain H37Ra.
Epitope: Extracellular domain

Application

Flow Cytometry Analysis: A representative lot detected α-galactosylceramide-/αGC-complexed CD1d on murine MH-S alveolar macrophages, LA-4 lung adenoma, and JAWSII bone marrow-derived dendritic cells (Rizvi, Z.A., et al. (2015). Toxicol. In Vitro. 29(6);1275-82.
Flow Cytometry Analysis: A representative lot detected αGC C20:2- and αGC26:0-complexed CD1d on the subpopulation of splenocytes with high CD11c-expression from mice injected with the respective α-GalCer analogs (Arora, P., et al. (2014). Immunity. 40(1):105-116).
Immunoprecipitation Analysis: A representative lot immunoprecipitated monoglycosylceramides-complexed mCD1d from rat RBL-Cd1d cells stably expressing murine CD1d (Kain, L., et al. (2014). Immunity. 41(4):543-554).
Immunocytochemistry Analysis: A representative lot detected α-GalCer-complexed murine CD1d on the surface of rat RBL-Cd1d cells stably expressing murine CD1d by fluorescent immunocytochemistry (Kain, L., et al. (2014). Immunity. 41(4):543-554).
Immunohistochemistry Analysis: A representative lot detected αGC C20:2- and αGC26:0-complexed CD1d on splenocytes in frozen spleen sections from mice injected with the respective α-GalCer analogs (Arora, P., et al. (2014). Immunity. 40(1):105-116).
ELISA Analysis: A representative lot selectively reacted with KRN7000-/α-GalCer-, OCH-, or C20:2-complexed mCD1d, but not glycolipid-free mCD1d. Clone L363 does not react with iGb3-complexed mCD1d, KRN7000-complexed hCD1b or KRN7000-complexed hCD1d (Yu, K.O., et al. (2007). J. Immunol. Methods. 323(1):11-23).
Neutralizing Analysis: A representative lot blocked CD1d-expressing cells from stimulating DN32.D3 NKT cells (Kain, L., et al. (2014). Immunity. 41(4):543-554).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-CD1d/KRN7000 Antibody, clone L363 is validated for use in Flow Cytometry, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry and ELISA for the detection of CD1d/KRN7000.

Quality

Evaluated by Flow Cytometry in alpha-GalCer glycolipid-treated RMA-S/mCD1d (WT) cells.

Flow Cytometry Analysis: 0.1 µg of this antibody detected CD1d/KRN7000 in alpha-GalCer glycolipid-treated RMA-S/mCD1d (WT) cells.

Target description

~38 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service