Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC777

Sigma-Aldrich

Anti-ING1, clone EPR6788, Rabbit Monoclonal Antibody

clone EPR6788, from rabbit

Synonym(s):

Inhibitor of growth protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR6788, monoclonal

species reactivity

rat, mouse, human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ING1(3621)

General description

Inhibitor of growth protein 1 (ING1) is a tumor suppressor protein that can induce cell growth arrest and apoptosis. It is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. ING1 is localized to the nucleus. Reduced expression of ING1 has been detected in various cancers including Squamous cell carcinoma of the head and neck.

Immunogen

Synthetic peptide corresponding to human ING1.

Application

Flow Cytometry Analysis: A 1:10-100 dilution from a representative lot detected ING1 in permeabilised NIH/3T3cells.

Produced in collaboration with Epitomics
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
This Anti-ING1, clone EPR6788, Rabbit Monoclonal Antibody is validated for use in Western Blotting and Flow Cytometry for the detection of ING1.

Quality

Evaluated by Western Blotting in HepG2, HeLa, MCF-7, Jurkat, C6, RAW264.7, PC-12, and NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected ING1 in HepG2, HeLa, MCF-7, Jurkat, C6, RAW264.7, PC-12, and NIH/3T3 cell lysates.

Target description

~47 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service