Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABC711

Sigma-Aldrich

Anti-ABCB5, clone 5H3C6 Antibody

clone 5H3C6, from mouse

Synonym(s):

ATP-binding cassette sub-family B member 5, ABCB5 P-gp, P-glycoprotein ABCB5, ABCB5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H3C6, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCB5(340273)

General description

ATP-binding cassette sub-family B member 5, also known as ABCB5 P-gp or P-glycoprotein ABCB5, and encoded by the gene name ABCB5, belongs to the ATP-binding cassette (ABC) transporter superfamily of integral membrane proteins. These proteins participate in ATP-dependent transmembrane transport of structurally diverse molecules ranging from small ions, sugars, and peptides to more complex organic molecules. ABCB5 is responsible for the resistance to doxorubicin of a subset of malignant melanomas and is expressed in several malignant tissues.

Immunogen

Purified recombinant fragment of human ABCB5 expressed in E. Coli

application

Immunofluorescence Analysis: A 1:200-1:1,000 dilution from a representative lot detected ABCB5 in HepG2 cells.

Flow Cytometry Analysis: A 1:200-1:400 dilution from a representative lot detected ABCB5 in HepG2 cells.

Immunohistochemistry Analysis: A 1:200-1:1,000 dilution from a representative lot detected ABCB5 in liver and breast cancer tissues.

ELISA: A representative lot was used on Control Antigen (100 ng) and Antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
This Anti-ABCB5, clone 5H3C6 Antibody is validated for use in western blotting, immunofluorescence, flow cytometry, IHC & ELISA for the detection of ABCB5.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:500-1:2,000 dilution of this antibody detected ABCB5 in A431 cell lysate.

Target description

~90 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing PBS with up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service