Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC702

Sigma-Aldrich

Anti-EPHB6, clone 8E7H12 Antibody

clone 8E7H12, from mouse

Synonym(s):

Ephrin type-B receptor 6, HEP, Tyrosine-protein kinase-defective receptor EPH-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8E7H12, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EPHB6(2051)

General description

Ephrin type-B receptor 6, also known as HEP or Tyrosine-protein kinase-defective receptor EPH-6, and encoded by the gene name EPHB6, belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. Eph receptors and ephrin ligands are membrane-bound cell-cell communication molecules with well-defined functions in development. EPHB6 is a unique member in the Eph family of receptor tyrosine kinases in that its kinase domain contains several alterations in conserved amino acids and is catalytically inactive. EPHB6 is expressed in brain and pancreas with lower levels found in heart, placenta, lung, liver, skeletal muscle, and kidney. EPHB6 can both positively and negatively regulate cell adhesion and migration and tyrosine phosphorylation of the receptor by an Src family kinase acts as the molecular switch for the functional transition.

Immunogen

Purified recombinant fragment of human EphB6 expressed in E. Coli

Application

Detect EPHB6 using this Anti-EPHB6, clone 8E7H12 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1:1,000 dilution from a representative lot detected EPHB6 in human bladder carcinoma tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers

Quality

Evaluated by Western Blotting in Jurkat and NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:500-1:2,000 dilution of this antibody detected EPHB6 in Jurkat and NIH/3T3 cell lysates.

Target description

~120 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service